Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Includes
- Primary Antibody
- NFκB Capture Probe
- NFκB Specific Competitor Oligonucleotide
- TFA Negative Control Probe
- HeLa Whole Cell Extract, TNFα Treated
- EZ-TFA (Universal Transcription Factor Assay, Chemiluminescent)
- Provided in a 96-well format
- When the Capture Probe, a double stranded biotinylated oligonucleotide is incubated together, the active form of NFkB contained in the nuclear extract binds to its consensus sequence
- After incubation, the extract/probe/buffer mixture is then directly transferred to the streptavidin coated plate
- The biotinylated double stranded oligonucleotide bound by active NFkB protein is immobilized and any inactive, unbound material is washed away
- The bound NFkB transcription factor subunits, p50 and p65, are detected with specific primary antibodies, a Rabbit anti-NFkB p50 (active form) and a Rabbit anti-NFkB p65
- An HRP-conjugated secondary antibody is then used for detection and provides sensitive colorimetric detection that can be read in a spectrophotometric plate reader
Specifications
Specifications
| Content And Storage |
|
| Detection Method | Chemiluminescence |
| For Use With (Application) | Affinity Binding Assay, ELISA, Activity Assay |
| Quantity | 1 Plate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?