Learn More
MilliporeSigma™ NFKB p65 EZ-TFA Transcription Factor Assay Colorimetric Kit
Catalog No. 70520 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Plate
Catalog No. Quantity
70-520 1 Plate
1 options
Catalog No. 70-520 Supplier MilliporeSigma™ Supplier No. 70520
Specifications
Content And Storage
  • Store kit components at the temperatures indicated on the labels and insert until their expiration date
  • Store HeLa Whole Cell Extract at -80°C. Avoid repeated freeze-thaw cycles. Stable at -80°C for 6 months
Quantity 1 Plate