Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NF-κB p65 (pS529) Mouse anti-Human, PE-CF594, Clone: K10-895.12.50, BD Phosflow™

Catalog No. BDB565447 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565447 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565447 Supplier BD Biosciences Supplier No. 565447
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

NF-κB p65 (pS529) Monoclonal antibody specifically detects NF-κB p65 (pS529) in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .
TRUSTED_SUSTAINABILITY

Specifications

Antigen NF-κB p65 (pS529)
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone K10-895.12.50
Conjugate PE-CF594
Description Vol. Per test: 5μL
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias RELA; REL-A; NFKBp65; NFKB3; Transcription factor p65; TF65
Host Species Mouse
Immunogen Phosphorylated Human NF-κB p65 Peptide
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.