Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NF-H Polyclonal Antibody
GREENER_CHOICE

Catalog No. APSA6753 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APSA6753 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APSA6753 Supplier Invitrogen™ Supplier No. PA316753
  • $625.00
    $494.65 / Each of 1

    Save $130.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chicken Polyclonal Antibody

NF-H Polyclonal Antibody for Western Blot, IHC (P), IHC (F), IHC (Free), ICC/IF, ELISA

Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NF-H
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Tissue culture supernatant with 0.02% sodium azide
Gene Nefh
Gene Accession No. P12036, P12037, P16884, P19246
Gene Alias 200 kDa neurofilament protein; CMT2CC; heavy neurofilament protein; I79_012512; intermediate filament protein; KIAA0845; LOW QUALITY PROTEIN: neurofilament heavy polypeptide; mKIAA0845; NEF3; Nefh; nefh.L; neurofilament; neurofilament 200kDa; neurofilament heavy; neurofilament heavy L homeolog; neurofilament heavy polypeptide; neurofilament heavy polypeptide-like; neurofilament triplet H protein; neurofilament, heavy polypeptide; neurofilament, heavy polypeptide 200kDa; neurofilament, heavy polypeptide L homeolog; neurofilament-H; NF 200 kD; NF200; Nfh; NF-H; NHC; XELAEV_18007418mg
Gene Symbols Nefh
Host Species Chicken
Immunogen Native NF-H (phosphorylated) purified from bovine spinal cord.
Purification Method IgY fraction
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100064328, 100156492, 101087272, 24587, 380684, 442940, 4744, 528842
Target Species Human, Mouse, Rat, Canine, Bovine, Pig, Feline, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.