Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NF-H Recombinant Superclonal™ Antibody (4HCLC)

Catalog No. 711025 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-102-5 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-102-5 Supplier Invitrogen™ Supplier No. 711025
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Superclonal Antibody

This antibody is predicted to react with Cat, Dog, Monkey and Bovine. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.

Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NF-H
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Recombinant Superclonal
Clone 4HCLC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene Nefh
Gene Accession No. P12036, P16884, P19246
Gene Alias 200 kDa neurofilament protein; CMT2CC; heavy neurofilament protein; I79_012512; intermediate filament protein; KIAA0845; LOW QUALITY PROTEIN: neurofilament heavy polypeptide; mKIAA0845; NEF3; Nefh; nefh.L; neurofilament; neurofilament 200kDa; neurofilament heavy; neurofilament heavy L homeolog; neurofilament heavy polypeptide; neurofilament heavy polypeptide-like; neurofilament triplet H protein; neurofilament, heavy polypeptide; neurofilament, heavy polypeptide 200kDa; neurofilament, heavy polypeptide L homeolog; neurofilament-H; NF 200 kD; NF200; Nfh; NF-H; NHC; XELAEV_18007418mg
Gene Symbols Nefh
Host Species Rabbit
Immunogen Peptide corresponding to Human NEFH (aa 631-643).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24587, 380684, 4744
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.