Learn More
Abcam NF-kappaB Activation Inhibitor VI, Benzoxathiole Compound, Solid, 5 mg
Description
- Molecular weight: 273.4 Da
- Purity: >95%
- Form: Solid
- Mechanism: Cell-permeable benzoxathiole compound that acts as an IKK-2 inhibitor, presumably by targeting the Ser177 and/or Ser181 residues in the kinase activation loop domain.
- Activity: Inhibits LPS-induced NF-κB activation (IC50 = 1.0 μM) and nitrite production (IC50 = 0.7 μM) in RAW 264.7 cells in vitro and effectively prevents LPS-induced septic death in mice in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | IKK-2 |
| Substrate Properties | Chemical Substrate |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Quantity | 5 mg |
| For Use With (Application) | NF-κB pathway inhibition, IKK-2 inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.