Learn More
Abcam NF-kappaB Activation Inhibitor VI, Benzoxathiole Compound, Solid, >95% Purity, 25 mg
Description
- Purity: >95%
- Molecular Weight: 273.4 Da
- Mechanism: Cell-permeable benzoxathiole compound that acts as an IKK-2 inhibitor
- Key Activity: Inhibits LPS-induced NF-κB activation (IC50 = 1.0 μM) and nitrite production (IC50 = 0.7 μM) in RAW 264.7 cells
- In Vivo Effect: Prevents LPS-induced septic death in mice
- Physical Form: Solid
- CAS Number: 113760-29-5
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | IKK-2 |
| Shipping Condition | Ambient |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Formulation | 100% NF-κB Activation Inhibitor VI |
| Quantity | 25 mg |
| For Use With (Application) | NF-κB pathway inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.