Learn More
Enzo Life Sciences NF-kappaB p65 polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADIKASTF110D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human p65 NF-κB. The sequence is completely conserved in mouse NF-κB p65. UniProt ID: Q04206. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Rabbit, Porcine, Monkey, Guinea pig, Dog, Bovine. Applications: WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7322
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.