Learn More
AdipoGen NF-kB (p65) (human) (rec.) (His) (highly active)
SDP
Supplier:  AdipoGen AG40T0020C002
Encompass_Preferred
NF-kB (p65) (human) (rec.) (His) (highly active), AdipoGen Life Sciences. Protein. Human NF-kappaB (p65) is fused to a His-tag. Source: Sf21 cells. Liquid. In 50mM TRIS-HCl, pH 7.5, containing 100mM sodium chloride, 0.2% NP-40, 50mM imidazole and 10% glycerol. Purity: >95% (SDS-PAGE). NF-kappaB is a pleiotropic transcription factor present in almost all cell types and is the endpoint of a series of signal transduction events that are initiated by a vast array of stimuli related to many biological processes such as inflammation, immunity, differentiation, cell growth, tumorigenesis and apoptosis. NF-kappaB is a homo- or heterodimeric complex formed by the Rel-like domain-containing proteins RELA/p65, RELB, NFKB1/p105, NFKB1/p50, REL and NFKB2/p52.
Catalog No. 50-594-501
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.