Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NF1 Monoclonal Antibody (McNFn27a)
GREENER_CHOICE

Catalog No. 01670103 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-103 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-103 Supplier Invitrogen™ Supplier No. MA1084
  • $579.89
    $449.65 / Each of 1

    Save $130.24

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA1-084 detects neurofibromin in human, rat and mouse cells. MA1-084 has been successfully used in Western blot, procedures. By Western blot, this antibody detects a ~230-260 kDa protein. The MA1-084 immunogen is a synthetic peptide corresponding to the N-terminal residues 27-41 of human neurofibromin.

The protein encoded by this gene is a type I integral membrane protein localized in the intermediate region between the endoplasmic reticulum and the Golgi, presumably recycling between the two compartments. The protein is a mannose-specific lectin and is a member of a novel family of plant lectin homologs in the secretory pathway of animal cells. Mutations in the gene are associated with a coagulation defect. Using positional cloning, the gene was identified as the disease gene leading to combined factor V-factor VIII deficiency, a rare, autosomal recessive disorder in which both coagulation factors V and VIII are diminished.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NF1
Applications Affinity Purification, ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone McNFn27a
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene NF1
Gene Accession No. P21359, P97526, Q04690
Gene Alias AW494271; DKFZp686J1293; E030030H24Rik; FLJ21220; neurofibromatosis 1; Neurofibromatosis type 1; neurofibromatosis-related protein NF-1; Neurofibromin; neurofibromin 1; Neurofibromin truncated; NF1; Nf-1; NFNS; VRNF; WSS
Gene Symbols NF1
Host Species Mouse
Immunogen Synthetic peptide corresponding to the N-terminal residues 27-41 of human neurofibromin.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18015, 24592, 4763
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.