Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFE2L1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501731358 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1358 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1358 Supplier Proteintech Group Inc Supplier No. 170621AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human ESRRG.

This gene encodes a protein that is involved in globin gene expression in erythrocytes. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene, NFE2L1, and for ′nuclear respiratory factor 1′ which has an official symbol of NRF1.

Specifications

Antigen NFE2L1
Applications Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene NFE2L1
Gene Accession No. Q14494, Q61985
Gene Alias HBZ17, LCR F1, Locus control region factor 1, NF E2 related factor 1, NFE2 related factor 1, NFE2L1, NRF1, TCF 11, TCF11, Transcription factor 11, Transcription factor HBZ17, Transcription factor LCR F1
Gene Symbols Nfe2l1
Host Species Rabbit
Immunogen NFE2L1 Fusion Protein Ag10832
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18023, 4779
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.