Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFkB p50 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 513500
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-350-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-350-0 Supplier Invitrogen™ Supplier No. 513500
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

NFkB p50 Polyclonal Antibody for ELISA, GS, ICC/IF, IHC (P), Western Blot

NFkB is a nuclear transcription factor activated by various extra and intracellular stimuli such as cytokines, UV radiation, stress, in injury, and by bacterial and viral products. NFkB is involved in regulation of various cellular events including cell growth, differentiation, proliferation, apoptosis and inflammation. NFKB1 (p50) is a 50KDa functional sub-unit of NF-kB and a member of Rel protein family which is synthesized as a p105 precursor protein and consists of an N-terminal conserved RHD-region containing nuclear localization signal, DNA-binding and dimerization domains. NFKB1 (p50) forms homodimers or heterodimerizes with p65, forming the functional NFkB factor. Further, NFKB1 (p50) directs the nuclear translocation of NF-kB and is instrumental in its DNA-binding. The NFkB complex is expressed in most cell types, and is primarily found in the cytoplasm in an inactive state in association with I-kappa-B. Phosphorylation of I-kappa-B releases the complex, allowing translocation to the nucleus and modification of gene expression. Phosphorylation of the p50 subunit at the Ser-337 residue has been shown to significantly increase DNA binding efficiency of the NFkB protein complex. The pathological role of NFkB has been suggested in AIDS, hematogenic cancer cell metastasis and rheumatoid arthritis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFkB p50
Applications ELISA, Gel Shift, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene NFKB1
Gene Accession No. P19838, P25799
Gene Alias CVID12; DKFZp686C01211; DNA-binding factor KBF1; EBP-1; KBF1; MGC54151; NF kappaB1; NF KB; NFkappaB; NF-kappaB; NF-kappa-B; NF-kappaB p50; NF-kappaB1; NF-kappa-B1 p84/NF-kappa-B1 p98; NF-kappabeta; NF-kB; NF-kB p50; NFKB1; NF-kB1; NFKB-p105; NFKB-p50; nuclear factor; nuclear factor kappa b; nuclear factor kappa B subunit 1; nuclear factor kappa-B 1; nuclear factor kappa-B DNA binding subunit; nuclear factor kappaB p50; nuclear factor NF-kappa-B p105 subunit; nuclear factor NF-kappa-B p105 subunit-like protein; nuclear factor NF-kappa-B p50 subunit; nuclear factor of kappa light chain gene enhancer in B-cells 1, p105; nuclear factor of kappa light polypeptide enhancer in B-cells 1; nuclear factor of kappa light polypeptide gene enhancer in B cells 1, p105; nuclear factor of kappa light polypeptide gene enhancer in B-cells 1; nuclear factor of kappa light polypeptide gene enhancer in B-cells 1 (p105); nuclear factor of kappa light polypeptide gene enhancer in B-cells 1, p105; nuclear factor-kappaB p50; OTTHUMP00000219572; p105; p50; p50 subunit of NF kappaB; p50 subunit of NF-kappaB; p50/p105; RHD of nuclear factor NF-kappa-B p105 subunit
Gene Symbols NFKB1
Host Species Rabbit
Immunogen Recombinant protein derived from an internal region of the human NF-kappa-B (p50) (nuclear factor of kappa-B).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18033, 4790
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.