Learn More
Enzo Life Sciences NF-kappaB p65 subunit (human) (12-317), (recombinant) (untagged) (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW99950050
Encompass_Preferred
Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 1X Dulbecco's Phosphate Buffered Saline (DPBS), pH 7.4, containing 5% glycerol. Source: Produced in E. coli BL21 (λDE3). Recombinant human NF-κB p65 subunit (12-317). Long Term Storage: -80°C.
Catalog No. 50-201-2345
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.