Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam NFkB p65 Transcription Factor Assay Kit, Chemiluminescent, 1x96 Tests

Catalog No. 502704338
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4338 1 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4338 Supplier Abcam Supplier No. AB2072211X96TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemiluminescent assay kit for detecting NFkB p65 in cell or tissue extracts with a 3 hr 30 min assay time.

  • Assay Time: 3 hr 30 min
  • Sensitivity: < 400 ng/well
  • Detection Method: Luminescent
  • Results Type: Semi-Quantitative
  • Sample Types: Nuclear extracts
  • Applications: Assay NFkB p65 Transcription Factor
  • Compatible Equipment: Microplate reader
  • Citation Count: 1

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule NFkB p65
Run Time 3 hr. 30 min.
No. of Reactions 96 Tests
For Use With (Equipment) Microplate Reader
Product Type Western Blot Kit
Detection Location In-Solution Detection
Format Kit
Detection Method Chemiluminescence
Sample Type Nuclear Extracts
Quantity 1 x 96 Tests
For Use With (Application) NFkB p65 Transcription Factor Assay
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.