Learn More
Abcam NFkB p65 Transcription Factor Assay Kit, Chemiluminescent, 5x96 Tests
Description
- Assay Time: 3 hr 30 min
- Detection Method: Luminescent
- Sensitivity: < 400 ng/well
- Results Type: Semi-Quantitative
- Sample Types: Nuclear Extracts
- Citation Count: 1
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | NFkB p65 |
| For Use With (Equipment) | Microplate Reader |
| Detection Method | Chemiluminescence |
| Product Type | Western Blot Kit |
| Detection Location | In-Solution Detection |
| Format | Kit |
| No. of Reactions | 480 Tests |
| Sample Type | Nuclear Extracts |
| Quantity | 5 x 96 Tests |
| For Use With (Application) | Transcription Factor Assay |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.