Learn More
Novus Biologicals™ NFkB Secreted Alkaline Phosphatase Reporter Assay Kit (Colorimetric)
Shop All R&D Systems Products

Description
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 1 Kit |
| Kit Contents | Secreted Placental Alkaline Phosphatase standard SEAP (200μg/ml), PNPP Substrate, pNF-KB/SEAP plasmid (0.5μg/μL), Sample Dilution Buffer (10X), PNPP buffer (10x), 96 well assay plates (Qty. 4), Plate sealer (Qty. 1), pCMV/SEAP plasmid (0.5μg/μL), 2 x 10μg |
| For Use With (Application) | ELISA |
| Product Type | NFkB Secreted Alkaline Phosphatase Reporter Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.