Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs NFPA Placard Kits

Catalog No. 18999806 Shop All AccuformNMC Products
Change view
Click to view available options
Dimensions (L x W):
10 x 10 in.
15 x 15 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
18-999-806 10 x 10 in.
18-999-807 15 x 15 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-999-806 Supplier Accuform Signs Supplier No. ZFD101EV
Add to Cart
Add to Cart

Includes

One NFPA placard and a full set of self-adhering number panels

Designed in accordance with standards set by NFPA for identifying potential hazards for firefighting crews

  • Ideal for mounting on or near containers of hazardous materials and equipment
  • Blank panels printed with the proper color coding using UV stable inks
  • Hazard panels printed on 4mil clear, self-adhesive polyester
  • Numbers and specific hazard symbols included (3 sets of 0-4 and six symbols)
  • Choice of placards made of either durable pressure-sensitive vinyl that mounts on most clean, dry surfaces, or high-gloss, 0.060 in. Dura-Plastic suitable for indoor/outdoor use

Specifications

Includes 1 NFPA placard and a full set of self-adhering number panels
Material Vinyl
Quantity 1 Ea.
For Use With (Application) Mounting on or near containers of hazardous materials and equipment
Height (English) 10 in.
Max. Temperature (English) 176°F
Min. Temperature (English) -20°F
Width (English) 10 in.
Temperature Range (Metric) -28°C to +80°C
Temperature Range (English) -20°F to +176°F
Dimensions (L x W) 10 x 10 in.
Product Type NFPA Placard Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.