Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFRKB Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7140892
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0450 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0450 Supplier Bethyl Laboratories, Inc Supplier No. A301459AT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Binds to the DNA consensus sequence 5′-GGGGAATCTCC-3′. Modulates the deubiquitinase activity of UCHL5.

Specifications

Antigen NFRKB
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene NFRKB
Gene Accession No. Q6P4R8
Gene Alias INO80G, DNA-binding protein R kappa B, DNA-binding protein R kappa-B, INO80 complex subunit G, nuclear factor related to kappa-B-binding protein
Gene Symbols NFRKB
Host Species Rabbit
Immunogen Between 1025 and 1075
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4798
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.