Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599074
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599074 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599074 Supplier Invitrogen™ Supplier No. PA599074
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NFS1 Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA

NFS1 (nitrogen fixation 1), also known as NIFS or IscS (cysteine desulfurase), is a member of the class V pyridoxal-phosphate-dependent aminotransferase family. It localizes to the cytoplasm or mitochondrion depending on which form is generated based on cytosolic pH. Highest expression levels of NFS1 are found in heart and skeletal muscle. Lower levels of expression are also found in liver, brain and pancreas. NFS1 is responsible for catalyzing the removal of sulfur from cysteine to form alanine, thereby supplying the inorganic sulfur for iron-sulfur (Fe-S) clusters. Fe-S clusters function as essential cofactors in a wide variety of events, including facilitation of electron transfer processes in oxidative phosphorylation, catalysis of enzymatic reactions in aconitase and dehydratases and maintenance of structural integrity in the DNA repair enzyme endonuclease III.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFS1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NFS1
Gene Accession No. Q9Y697
Gene Alias AA987187; cysteine desulfurase, mitochondrial; Cysteine desulfurase, mitochondrial-like protein; HUSSY-08; IscS; LOW QUALITY PROTEIN: cysteine desulfurase, mitochondrial; m-Nfs1; m-Nfsl; Nfs1; NFS1 cysteine desulfurase; NFS1 nitrogen fixation 1 homolog; NFS1 nitrogen fixation 1 homolog (S. cerevisiae); NFS1, cysteine desulfurase; Nifs; nifS-like (sic); nitrogen fixation 1 (S. cerevisiae, homolog); nitrogen fixation gene 1; nitrogen fixation gene 1 (S. cerevisiae); nitrogen fixation gene, yeast homolog 1; nitrogen-fixing bacteria S-like protein
Gene Symbols NFS1
Host Species Rabbit
Immunogen Recombinant Human Cysteine desulfurase, mitochondrial protein (208-457AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9054
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.