Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFYB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531913 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531913 Supplier Invitrogen™ Supplier No. PA531913
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, HeLa nuclear extract, Neuro 2A, C8D30, NIH-3T3, Raw264.7, C2C12, PC-12, Rat2. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (92%), Pig (99%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The CCAAT-binding factor NF-Y is a heteromeric transcription factor that specifically binds to CCAAT sequences in many eukaryotic genes. NF-Y is made up of three subunits, NF-YA, NF-YB and NF-YC. All three components are necessary for DNA binding. In each NF-Y subunit, the segment needed for formation of the NF-Y-DNA complex is conserved from yeast to human. These conserved segments are homologous to the histone-fold motif of eukaryotic histones. The DNA-binding domains of the NF-YB and NF-YC subunits have been suggested to interact through a protein-protein, histone-fold 'handshake' motif in a manner analogous to the histone proteins H2B and H2A, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFYB
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene NFYB
Gene Accession No. P25208, P63139, P63140
Gene Alias AA985999; CAAT box DNA-binding protein subunit B; CAAT-box DNA-binding protein subunit B; CBF-A; CBFB; CBF-B; CCAAT binding transcription factor of CBF-B/NFY-B; CCAAT-binding transcription factor subunit A; CCAAT-binding transcription factor subunit B; HAP3; nf-y; Nfya; Nfyb; NF-YB; nuclear transcription factor - Y beta; nuclear transcription factor Y subunit B; Nuclear transcription factor Y subunit beta; nuclear transcription factor Y, beta; nuclear transcription factor-Y beta; Transcription factor NF-Y, B subunit
Gene Symbols NFYB
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NFYB. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18045, 25336, 4801
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.