Learn More
Enzo Life Sciences NG-Hydroxy-L-arginine . monoacetate (5mg). CAS: 53598-01-9
SDP
Supplier:  Enzo Life Sciences ALX106004M005
Encompass_Preferred
Intermediate in the biosynthesis of nitric oxide from L-arginine by nitric oxide synthases (NOS). Inhibitor of arginases. Alternative name: L-HOArg . AcOH. Formula: C6H14N4O3 . CH3COOH. MW: 190.2 . 60.1. Purity: ≥98% (HPLC). Solubility: Soluble in water (50mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-7734
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.