Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ NG-Test™ CARBA 5
SDP

Catalog No. 23001741 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Detection Method:
Rapid Detection
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Method
23-001-741 Rapid Detection
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-741 Supplier Hardy Diagnostics™ Supplier No. CARBA5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

NG-Test CARBA 5 is the only rapid, multiplex, phenotypic test capable of detecting KPC, OXA-48-like, VIM, IMP, and NDM carbapenemases produced by Enterobacterales and P. aeruginosa.

  • In vitro rapid and visual multiplex immunochromatographic assay for the qualitative detection and differentiation of five common carbapenemases
  • Intended as an aid for infection control in the detection of carbapenemase-producing Enterobacterales and Pseudomonas aeruginosa in healthcare settings
  • Cost-Effective:more economical than molecular methods
  • Accurate:unlike molecular methods, detects the gene expression which can be crucial to aiding therapeutic decisions
  • 5 simple step procedure

Specifications

Sample Type Environmental Sample, Food Sample
Target Organism Class E. coli, Total Coliforms, Fecal Coliforms, Enterococcus
Content And Storage Store devices in sealed pouches between 4°C and 30°C
Detection Method Rapid Detection
Target Molecule Carbapenemases
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.