Learn More
Alpco Ltd NGAL (Lipocalin-2) ELISA
SDP
Supplier:  Alpco Ltd 43NGALHUE01
Encompass_Preferred
The NGAL (Lipocalin-2) ELISA is a quantitative immunoassay for the determination of neutrophil gelatinase-associated lipocalin (NGAL) in human stool and urine. For research use only. Compatible with product 43-NGALEX-50 for NGAL stool extraction.
Catalog No. NC2885459
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.