Learn More
Alpco Ltd NGAL Sample Collection Kit
SDP
Supplier:  Alpco Ltd 43NGALEX50
Encompass_Preferred
The NGAL Sample Collection Kit includes 50 pre-filled tubes designed for extraction of stool samples for analysis in the NGAL (Lipocalin-2) ELISA, product # 43-NGALHU-E01.
Catalog No. NC2885460
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.