Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NGEF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143696
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143696 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143696 Supplier Invitrogen™ Supplier No. PA5143696
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects an 80 kDa* band corresponding to ephexin-1 in Western blots of human A431, Jurkat, and HUVEC cells, as well as rat PC12 cells and mouse brain tissue.

Acts as a guanine nucleotide exchange factor (GEF) which differentially activates the GTPases RHOA, RAC1 and CDC42. Plays a role in axon guidance regulating ephrin-induced growth cone collapse and dendritic spine morphogenesis. Upon activation by ephrin through EPHA4, the GEF activity switches toward RHOA resulting in its activation. Activated RHOA promotes cone retraction at the expense of RAC1- and CDC42-stimulated growth cone extension.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NGEF
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene NGEF
Gene Accession No. Q5BKC9, Q8CHT1, Q8N5V2
Gene Alias ARHGEF27; Besh3; brain-enriched SH3-domain protein Besh3; ephexin; ephexin-1; eph-interacting exchange protein; Neuronal guanine nucleotide exchange factor; Ngef; Tims2; transforming immortalised mammary epithelial sequence 2
Gene Symbols NGEF
Host Species Rabbit
Immunogen Ephexin-1 synthetic peptide (coupled to carrier protein) corresponds to amino acids in the C-terminal region of mouse ephexin-1. This sequence has significant homology to the same region in human and rat ephexin-1, and has low homology to other known proteins.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246217, 25791, 53972
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.