Learn More
Invitrogen™ NGF Polyclonal Antibody

Description
PA1-9516 detects Nerve growth factor in human, mouse and rat samples. PA1-9516 has been successfully used in Western blot, Immunohistochemistry and ELISA procedures. The PA1-9516 immunogen is a synthetic peptides corresponding to residues 74-85 & 113-124 of mouse NGF protein.
Specifications
Specifications
| Antigen | NGF |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | NGF |
| Gene Accession No. | P01138, P01139, P25427 |
| Gene Alias | B nerve growth factor; beta ngf; beta-nerve growth factor; BetaNGF; beta-NGF; bNGF; b-NGF; HSAN5; MGC161426; MGC161428; Nerve growth factor; nerve growth factor (beta polypeptide); nerve growth factor, beta; nerve growth factor, beta polypeptide; nerve growth factor, beta subunit; NGF; NGF b; NGF β; Ngfb; NGFbeta; NGFβ; β nerve growth factor; β NGF; βNGF |
| Gene Symbols | NGF |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.