Learn More
Invitrogen™ NGF Polyclonal Antibody

Description
Reconstitute in 500 μL of sterile, ultrapure water. Centrifuge to remove any insoluble material. Store lyophilized antibody at 2-8 °C. After reconstitution keep aliquots at -20 °C to -80 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Avoid repetitive freeze/thaw cycles. Glycerol (1:1) may be added for an additional stability.
Specifications
Specifications
| Antigen | NGF |
| Applications | Inhibition Assays, Immunohistochemistry (Frozen), Neutralization, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | NGF |
| Gene Accession No. | P01138, P01139, P25427 |
| Gene Alias | B nerve growth factor; beta ngf; beta-nerve growth factor; BetaNGF; beta-NGF; bNGF; b-NGF; HSAN5; MGC161426; MGC161428; Nerve growth factor; nerve growth factor (beta polypeptide); nerve growth factor, beta; nerve growth factor, beta polypeptide; nerve growth factor, beta subunit; NGF; NGF b; NGF β; Ngfb; NGFbeta; NGFβ; β nerve growth factor; β NGF; βNGF |
| Gene Symbols | NGF |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.