Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NGF Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01670631
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-631 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-631 Supplier Invitrogen™ Supplier No. PA118379
Only null left
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Reconstitute in 250 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Avoid repetitive freeze/thaw cycles. Glycerol (1:1) may be added for an additional stability.

Nerve growth factor (NGF) influences the survival and differentiation of a variety of neuronal and nonneuronal cells. The actions of this polypeptide result from binding to specific cell surface receptors which are present as both high and low affinity sites. Although both types of receptor bind NGF only the high affinity form is capable of signal transduction. Cross-linking studies with 125I-NGF have shown that the high affinity receptor of rat PC12 cells forms an Mr 158,000 complex with NGF while the low affinity receptor forms an Mr 100,000 complex. Full length cDNA clones for both the human and rat NGF receptor genes have been produced and sequenced, revealing greater than 90% homology between the two species. NGF exhibits nerve growth stimulating activity and is involved in the regulation of growth and the differentiation of sympathetic and certain sensory neurons. Further, NGF stimulates division and differentiation of sympathetic and embryonic sensory neurons. Mutations in the NGF gene have been associated with hereditary sensory and autonomic neuropathy, type 5 (HSAN5), and dysregulation of NGF gene expression is associated with allergic rhinitis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NGF
Applications ELISA, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with no preservative
Gene NGF
Gene Accession No. P01138, P01139, P25427
Gene Alias B nerve growth factor; beta ngf; beta-nerve growth factor; BetaNGF; beta-NGF; bNGF; b-NGF; HSAN5; MGC161426; MGC161428; Nerve growth factor; nerve growth factor (beta polypeptide); nerve growth factor, beta; nerve growth factor, beta polypeptide; nerve growth factor, beta subunit; NGF; NGF b; NGF β; Ngfb; NGFbeta; NGFβ; β nerve growth factor; β NGF; βNGF
Gene Symbols NGF
Host Species Sheep
Immunogen Native mouse beta NGF purified from submaxillary salivary gland.
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18049, 310738, 4803
Target Species Human, Mouse, Rat, Avian
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.