Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NGFR Monoclonal Antibody (MC192)
GREENER_CHOICE

Catalog No. MA118417
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA118417 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA118417 Supplier Invitrogen™ Supplier No. MA118417
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. The MC192 is supplied in lyophilised form from Protein G-purified hybridoma cell culture supernatants. The lyophilised antibody is stable when stored at 2-8 °C or -20 °C. After reconstitution undiluted aliquots should be kept at -20 °C for up to six months. For additional stability Glycerol (1:1) may be added after reconstitution. Repetitive freeze/thaw cycle should be avoided.

Neurotrophic factors control the survival, differentiation and maintenance of neurons in the peripheral and central nervous systems, and of other neural crest-derived cell types. Developing sympathetic neurons are absolutely dependent upon nerve growth factor (NGF) during the period of target competition in vivo. Antibodies reacting specifically with NGFR p75 are useful tools in the detection and characterization of NGFR p75, to enhance our understanding of a wide range of phenomena in the development, plasticity and repair of the nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NGFR
Applications ELISA, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone MC192
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Ngfr
Gene Accession No. P07174
Gene Alias CD271; CD271 antigen; Gp80-LNGFR; LNGFR; low affinity nerve growth factor receptor; low affinity neurotrophin receptor p75NTR; LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 16; low-affinity nerve growth factor receptor; nerve growth factor receptor; nerve growth factor receptor (TNFR superfamily, member 16); nerve growth factor receptor, fast; NGF receptor; NGFR; NGR; p75; p-75; p75 ICD; p75 neurotrophin receptor; p75 ntr; p75(NTR); p75NGFR; p75NTR; RNNGFRR; TNFR superfamily, member 16; Tnfrsf16; TNR16; Tumor necrosis factor receptor superfamily member 16
Gene Symbols Ngfr
Host Species Mouse
Immunogen NGF receptor (aa 30-425).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24596
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.