Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NGFR Monoclonal Antibody (ME20.4)
GREENER_CHOICE

Catalog No. PIMA120167 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA120167 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA120167 Supplier Invitrogen™ Supplier No. MA120167
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Recommended positive controls: human tongue. This antibody does not cross-react with mouse, rat, or chicken p75 NGF receptors. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Neurotrophic factors control the survival, differentiation and maintenance of neurons in the peripheral and central nervous systems, and of other neural crest-derived cell types. Developing sympathetic neurons are absolutely dependent upon nerve growth factor (NGF) during the period of target competition in vivo. Antibodies reacting specifically with NGFR p75 are useful tools in the detection and characterization of NGFR p75, to enhance our understanding of a wide range of phenomena in the development, plasticity and repair of the nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NGFR
Applications ELISA, Flow Cytometry, Inhibition Assays, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone ME20.4
Concentration 2.8 mg/mL
Conjugate Unconjugated
Formulation Ascites with 15mM sodium azide
Gene Ngfr
Gene Accession No. P08138
Gene Alias CD271; CD271 antigen; Gp80-LNGFR; LNGFR; low affinity nerve growth factor receptor; low affinity neurotrophin receptor p75NTR; LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 16; low-affinity nerve growth factor receptor; nerve growth factor receptor; nerve growth factor receptor (TNFR superfamily, member 16); nerve growth factor receptor, fast; NGF receptor; NGFR; NGR; p75; p-75; p75 ICD; p75 neurotrophin receptor; p75 ntr; p75(NTR); p75NGFR; p75NTR; RNNGFRR; TNFR superfamily, member 16; Tnfrsf16; TNR16; Tumor necrosis factor receptor superfamily member 16
Gene Symbols Ngfr
Host Species Mouse
Immunogen Human melanoma cell line WM245.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100525607, 101101519, 4804
Target Species Feline, Human, Monkey, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.