Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NGLY1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571345
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571345 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571345 Supplier Invitrogen™ Supplier No. PA571345
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NGLY1 Polyclonal Antibody for Western Blot

This gene encodes an enzyme that catalyzes hydrolysis of an N(4)-(acetyl-beta-D-glucosaminyl) asparagine residue to N-acetyl-beta-D-glucosaminylamine and a peptide containing an aspartate residue. The encoded enzyme may play a role in the proteasome-mediated degradation of misfolded glycoproteins. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NGLY1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NGLY1
Gene Accession No. Q96IV0
Gene Alias 1110002C09Rik; CDDG; CDG1V; hPNGase; mPNGase; NGLY1; N-glycanase 1; peptide N-glycanase homolog; Peptide:N-glycanase; peptide-N(4)-(N-acetyl-beta-glucosaminyl)asparagine amidase; PNG1; PNGase
Gene Symbols NGLY1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human NGLY1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55768
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.