Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NgR3/NgRH2/RTN4RL1 Antibody (319123) [DyLight 405], R&D Systems™
SDP

Catalog No. FAB2920E
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2920E 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB2920E Supplier R&D Systems Supplier No. FAB2920E

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

NgR3/NgRH2/RTN4RL1 Monoclonal antibody specifically detects NgR3/NgRH2/RTN4RL1 in Human samples. It is validated for Western Blot

Specifications

Antigen NgR3/NgRH2/RTN4RL1
Applications Western Blot
Classification Monoclonal
Clone 319123
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias DKFZp547J144, NgR3, NGRH2, NGRL2, nogo receptor-like 2, Nogo-66 receptor homolog 2, Nogo-66 receptor-related protein 3, reticulon 4 receptor-like 1, reticulon-4 receptor-like 1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human NgR3/NgRH2, Cys25-Ala419, Accession # Q86UN2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146760
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.