Learn More
Medchemexpress LLC HY-42427 100mg Medchemexpress, NH2-PEG2-CH2-Boc CAS:1948273-09-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY42427100MG
Encompass_Preferred
Medchemexpress, HY-42427 100mg NH2-PEG2-CH2-Boc CAS:1948273-09-3 NH2-PEG2-CH2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2241
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.