Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NHE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583345
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583345 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583345 Supplier Invitrogen™ Supplier No. PA583345
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DNPVFSPDEA LDRSLLARLP PWLSPGETVV PSQRARTQIP YSPGTFCRLM PFRLSSKSVD SFLQADGPEE RPPAALPEST Highest antigen sequence indentity to the following orthologs: Mouse - 78%, Rat - 79%.

NHE3 (also known as SLC9A3 - Sodium/hydrogen exchanger 3) is a member of the sodium/hydrogen exchanger family that plays a crucial role in acid-base and volume homeostasis by mediating the majority of sodium and bicarbonate reabsorption in the proximal tubule of the kidney. Mutations in the gene can lead to Diarrhea 8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NHE3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC9A3
Gene Accession No. P48764
Gene Alias 9030624O13Rik; AI930210; DIAR8; MGC126718; MGC126720; Na(+)/H(+) exchanger 3; Na/H exchanger 3; Na+/H+ exchanger homolog; Na+/H+ exchanger isoform 3; membrane ion exchange protein; Nhe3; NHE-3; NHE-3 antibody; plasma membrane ion transport protein; putative sodium hydrogen exchanger subtype 3; Slc9a3; SLC9A3 antibody; sodium proton exchanger; sodium/hydrogen exchanger; sodium/hydrogen exchanger 3; sodium/hydrogen exchanger isoform 3; Solute carrier family 9 (sodium/hydrogen exchanger 3), antiporter 3, Na+/H+ (amiloride insensitive); solute carrier family 9 (sodium/hydrogen exchanger), isoform 3; solute carrier family 9 (sodium/hydrogen exchanger), member 3; solute carrier family 9 member 3; solute carrier family 9 member A3; solute carrier family 9, member 3; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3
Gene Symbols SLC9A3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC9A3. Recombinant protein control fragment (Product #RP-96810).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6550
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.