Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NHLRC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140347 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140347 Supplier Invitrogen™ Supplier No. PA5140347
  • $523.00
    $457.65 / Each of 1

    Save $65.35

    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VGPGPDGGLA VSEEFGDVRL FGSARQPLGS LGGWTGHTFG CPAGICSNSE GNVIVVDEQ Highest antigen sequence indentity to the following orthologs: Mouse - 78%, Rat - 76%.

NHLRC4 (NHL Repeat Containing 4) is a Protein Coding gene. An important paralog of this gene is PRR35. Predicted to enable ubiquitin protein ligase activity. Predicted to be involved in protein K48-linked ubiquitination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NHLRC4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NHLRC4
Gene Accession No. P0CG21
Gene Alias NHL Repeat Containing 4; NHL Repeat-Containing Protein 4
Gene Symbols NHLRC4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NHLRC4. Recombinant protein control fragment (Product #RP-109624).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283948
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.