Learn More
R&D Systems NHP XL CXCL2/GRO beta, Magnetic Luminex™ Performance Assay
Shop All Novus Biologicals Products

Specifications
Specifications
| Quantity | 1 Kit |
| Species | Primate |
| Binding Target | CXCL2/GRO beta/MIP-2/CINC-3 |
| Gene ID (Entrez) | 2920 |
| Product Line | R&D Systems |
| Synonym | chemokine (C-X-C motif) ligand 2, CINC-2a, CINC3, CINC-3, C-X-C Motif Chemokine 2, GRO beta, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MGSA-beta, MIP2, MIP-2, MIP-2a, MIP2A, MIP2-alpha, SCYB2 |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.