Learn More
R&D Systems NHP XL IFN-gamma, Magnetic Luminex™ Performance Assay
Shop All Novus Biologicals Products

Specifications
Specifications
| Quantity | 1 Kit |
| Species | Primate |
| Binding Target | IFN-gamma |
| Gene ID (Entrez) | 3458 |
| Product Line | R&D Systems |
| Synonym | IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.