Learn More
Sigma Organic Chemistry NHS-dPEG(R)4-biotin | 50MG | 459426-22-3
Supplier: Sigma Organic Chemistry QBD1020050MG

NHS-dPEG(R)4-biotin, 50MG
About This Item:
Linear Formula: C25H40N4O10S
UNSPSC Code: 12352200
Storage: -20C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.