Learn More
Medchemexpress LLC NHS-modified MMAF TFA | 96.0% | 1165.34 g/mol | C57H87F3N8O14 | 5 MG

Supplier: Medchemexpress LLC HY139325A5MG
NHS-modified MMAF TFA is a trifluoroacetate (TFA) salt intermediate used in the synthesis of antibody-drug conjugates (ADCs), notably for preparing anti-mesothelin binder-agent conjugates.
- Purity 96.03%
- Molecular weight 1165.34 g/mol
- Chemical formula C57H87F3N8O14
- Physical state: white to off-white solid
- Available package sizes: 1 mg, 5 mg, 10 mg, 50 mg, 100 mg
- Solubility in DMSO: 16 mg/mL (13.73 mM); sonication assisted
- Storage: -20°C protected from light; in solvent store -80°C (6 months) or -20°C (1 month)
- Available documentation: data sheet, SDS, COA, HNMR, RP-HPLC, LCMS, product usage guide
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.