Learn More
Sartorius Biosensor / Ni-NTA (NTA) Tray
Supplier:  Sartorius 185101
Encompass
Ni-NTA (NTA) Biosensors - One tray of 96 biosensors coated with nickel-charged tris-NTA for both quantitation and kinetic applications of HIS-tagged molecules.
Catalog No. NC0295617
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov