Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Ni Sepharose™ excel
GSA_VA

Catalog No. 45003012 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-003-012 500 mL
45-002-820 25 mL
45-003-011 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-003-012 Supplier Cytiva Supplier No. 17371203
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Ni Sepharose excel affinity media for capture and purification of histidine-tagged proteins secreted into eukaryotic cell culture supernatants by immobilized metal ion affinity chromatography (IMAC).

Ni Sepharose excel enables direct loading of samples without having to perform extensive and time-consuming pretreatment procedures.

  • Load eukaryotic cell culture samples containing secreted histidine-tagged proteins directly with retained binding capacity.
  • Increase target protein yield and decrease degradation through reduced and simplified sample handling.
  • Choose between several formats for screening and preparative purification of histidine-tagged proteins.
  • Ni Sepharose excel is an IMAC medium precharged with nickel ions very strongly bound to a chelating ligand, allowing loading of samples that usually cause stripping of metal ions.
  • Ni Sepharose excel is designed primarily for capture and purification of histidine-tagged proteins secreted into cell culture supernatants from eukaryotic cells such as insect cells or CHO cells.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Buffer 0.01M HCl and 0.01M NaOH (one week), 10mM EDTA, 5mM DTT, 1M NaOH, 5mM TCEP, 20mM beta-Mercapto ethanol and 6M Guanidine-HCl (24 Hours), 500mM Imidazole and 100mM EDTA (2 Hours), 30% 2-Propanol
Product Type Ni Sepharose excel chromato graphy media
Quantity 500 mL
Flow Rate 150 to 600cm/hr.
pH Range 2 to 12
Particle Size 90μm
Support Type 6% Highly Cross-linked Agarose
Content And Storage 4° to 30°C
For Use With (Application) Capture and purification of His-tagged Proteins secreted into eukaryotic cell culture supernatants by immobilized Metal ion affinity chromato graphy (IMAC)
Matrix Ni-Sepharose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.