Learn More
Medchemexpress LLC Nicotinic acid-d4 | 66148-15-0 | 99.2% | 127.13 g/mol | C6HD4NO2 | 10 MG
Supplier: Medchemexpress LLC HYB0143S210MG
Niacin-d4 (nicotinic acid-d4) is a deuterium-labeled stable isotope of niacin (vitamin B3) supplied as a calibrated reference material for analytical and metabolic studies. It is formulated for use as an internal standard or tracer in mass spectrometry, quantitative assays, and isotope-tracing experiments, and is supplied with reported high purity and recommended storage conditions.
- High isotopic labeling for accurate mass spectrometry quantification.
- Reported high purity (≈99.2%) suitable for analytical reference use.
- Supplied in small measured quantities suitable for trace-level studies.
- Solid form recommended stored sealed, protected from moisture and light at 4°C; in solution, store frozen.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.