Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niacinamide, FCC, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18604790 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
5 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-790 5 kg Poly Pail
18-604-787 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-604-790 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N10825KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N1082, 98-92-0, C6H6N2O

Spectrum™ Chemical Niacinamide, FCC is a vitamin supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 98-92-0
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
InChI Key DFPAKSUCGFBDDF-UHFFFAOYSA-N
IUPAC Name pyridine-3-carboxamide
SMILES NC(=O)C1=CC=CN=C1

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Melting Range 128-131°C
Packaging Poly Pail
Quantity 5 kg
Percent Purity 98.5 to 101%
Grade FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.