CAS RN 98-92-0
Thermo Scientific Chemicals Nicotinamide, 99%
CAS: 98-92-0 Molecular Formula: C6H6N2O Molecular Weight (g/mol): 122.13 MDL Number: MFCD00006395 InChI Key: DFPAKSUCGFBDDF-UHFFFAOYSA-N Synonym: nicotinamide,niacinamide,3-pyridinecarboxamide,vitamin pp,nicotinic acid amide,papulex,aminicotin,amixicotyn,nicobion,nicotylamide PubChem CID: 936 ChEBI: CHEBI:17154 IUPAC Name: pyridine-3-carboxamide SMILES: C1=CC(=CN=C1)C(=O)N
Nicotinamide 99.0+%, TCI America™
CAS: 98-92-0 Molecular Formula: C6H6N2O Molecular Weight (g/mol): 122.127 MDL Number: MFCD00006395 InChI Key: DFPAKSUCGFBDDF-UHFFFAOYSA-N Synonym: nicotinamide,niacinamide,3-pyridinecarboxamide,vitamin pp,nicotinic acid amide,papulex,aminicotin,amixicotyn,nicobion,nicotylamide PubChem CID: 936 ChEBI: CHEBI:17154 IUPAC Name: pyridine-3-carboxamide SMILES: C1=CC(=CN=C1)C(=O)N
R&D Systems™ Nicotinamide
A Tocris product, part of R&D Systems. Nicotinamide synthesized to Ancillary Material Grade
Nicotinamide, 99%
CAS: 98-92-0 Molecular Formula: C6H6N2O Molecular Weight (g/mol): 122.127 MDL Number: MFCD00006395 InChI Key: DFPAKSUCGFBDDF-UHFFFAOYSA-N Synonym: nicotinamide,niacinamide,3-pyridinecarboxamide,vitamin pp,nicotinic acid amide,papulex,aminicotin,amixicotyn,nicobion,nicotylamide PubChem CID: 936 ChEBI: CHEBI:17154 IUPAC Name: pyridine-3-carboxamide SMILES: C1=CC(=CN=C1)C(=O)N
Niacinamide, FCC, 98.5-101%, Spectrum™ Chemical
CAS: 98-92-0 Molecular Formula: C6H6N2O Molecular Weight (g/mol): 122.13 InChI Key: DFPAKSUCGFBDDF-UHFFFAOYSA-N IUPAC Name: pyridine-3-carboxamide SMILES: NC(=O)C1=CC=CN=C1
Medisca Niacinamide, USP (Nicotinamide) (Dietary Supplement Grade)
Niacinamide, USP (Nicotinamide) (Dietary Supplement Grade) is a white crystalline powder that is odorless or practically so.
R&D Systems™ Nicotinamide
CAS: 98-92-0 Molecular Formula: C6H6N2O
Vitamins - IR Identification PT, MilliporeSigma™ Supelco™
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
Niacinamide, USP, 98.5-101.5%, Spectrum™ Chemical
CAS: 98-92-0 Molecular Formula: C6H6N2O Molecular Weight (g/mol): 122.13 InChI Key: DFPAKSUCGFBDDF-UHFFFAOYSA-N IUPAC Name: pyridine-3-carboxamide SMILES: NC(=O)C1=CC=CN=C1
Niacinamide, USP, 98.5-101.5%, Spectrum™ Chemical
CAS: 98-92-0 Molecular Formula: C6H6N2O Molecular Weight (g/mol): 122.13 InChI Key: DFPAKSUCGFBDDF-UHFFFAOYSA-N IUPAC Name: pyridine-3-carboxamide SMILES: NC(=O)C1=CC=CN=C1