Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIBL Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7140804
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0130 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0130 Supplier Bethyl Laboratories, Inc Supplier No. A301338AT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat

The Niban-like protein is encoded by a maternal imprinting gene. This protein may possess growth suppressor activity. Two major alternatively spliced isoforms have been described. forms. One form, Meg3-proximal (Meg3p), contains exons 1-3. The second form, Meg3-distal (Meg3d) did not contain exons 1-3 and was present in oocytes and in 1- and 2-cell embryos. Adachi H, Majima S, et al. Oncogene. 2004 Apr 22;23(19):3495-500 Matsumoto F, Fujii H, et al. Hum Pathol. 2006 Dec; 37(12):1592-600.

Specifications

Antigen NIBL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene FAM129B
Gene Accession No. Q96TA1
Gene Alias C9orf88
Gene Symbols FAM129B
Host Species Rabbit
Immunogen Between 683 and 733
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64855
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.