Learn More
TARGETMOL CHEMICALS INC NIBR189 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T715610MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. NIBR189 is a potent and selective antagonist of EBI2 (GPR183) receptor (IC50 of 11 and 15 nM for human and mouse EBI2 receptors, respectively)
Catalog No. 50-287-8204
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.