Learn More
Selleck Chemical LLC Nicardipine HCl S4181-50mg
SDP
Supplier:  Selleck Chemical LLC S418150MG
Encompass_Preferred
Nicardipine HCl (RS-69216 YC-93) is a dihydropyridine calcium-channel blocking agent used for the treatment of vascular disorders
Catalog No. 50-311-1219
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.