Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nicastrin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1758 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1758 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1758 Supplier Invitrogen™ Supplier No. PA1758
  • $576.80
    $449.65 / Each of 1

    Save $127.15

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-758 detects nicastrin from human, mouse and rat samples. PA1-758 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~110 kDa protein representing nicastrin. PA1-758 immunizing peptide corresponds to amino acid residues 688-708 from human nicastrin. There is a single amino acid substitution between the human and mouse sequence. PA1-758 immunizing peptide (Cat. # PEP-131) is available for use in neutralization and control experiments.

NCSTN (Nicastrin) is a gene that encodes a type I transmembrane glycoprotein that is an integral component of the multimeric gamma-secretase complex. The encoded protein cleaves integral membrane proteins, including Notch receptors and beta-amyloid precursor protein, and may be a stabilizing cofactor required for gamma-secretase complex assembly. The cleavage of beta-amyloid precursor protein yields amyloid beta peptide, the main component of the neuritic plaque and the hallmark lesion in the brains of patients with Alzheimer's disease; however, the nature of the encoded protein's role in Alzheimer's disease is not known for certain. Mutations in this gene are associated with familial acne inversa. A pseudogene of this gene is present on chromosome 21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nicastrin
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene NCSTN
Gene Accession No. P57716, Q8CGU6, Q92542
Gene Alias 9430068N19Rik; AA727311; anterior pharynx-defective 2; Aph2; ATAG1874; D1Dau13e; Kiaa0253; mKIAA0253; NCSTN; Nct; Nicastrin; OTTHUMP00000031840; UNQ1874/PRO4317
Gene Symbols NCSTN
Host Species Rabbit
Immunogen Synthetic Peptide: C(688) I N A K A D V L F I A P R E P G A V S Y(708).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23385, 289231, 59287
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.