Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicergoline

Catalog No. AAJ6329503
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6329503 1 g
AAJ63295MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6329503 Supplier Thermo Scientific Chemicals Supplier No. J6329503
Add to Cart

CAS: 27848-84-6 | C24H26BrN3O3 | 484.394 g/mol

Nicergoline is an ergot derivative and vasodilator which increases blood flow in the brain. Nicergoline inhibits the vasoconstrictor effect of catecholamines by inhibiting the postsynaptic α1-adrenoceptors on vascular smooth muscle.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in alcohol, chloroform, and acetone. Insoluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27848-84-6
Molecular Formula C24H26BrN3O3
Molecular Weight (g/mol) 484.394
MDL Number MFCD00869626
InChI Key YSEXMKHXIOCEJA-FVFQAYNVSA-N
Synonym nicergoline, nicotergoline, nimergoline, nimergoline base, nicergolinum, nicergolin, nicergolina, sermion, nicergolin german, nicergolina dcit
PubChem CID 34040
IUPAC Name [(6aR,9R,10aS)-10a-methoxy-4,7-dimethyl-6a,8,9,10-tetrahydro-6H-indolo[4,3-fg]quinoline-9-yl]methyl 5-bromopyridine-3-carboxylate
SMILES CN1CC(CC2(C1CC3=CN(C4=CC=CC2=C34)C)OC)COC(=O)C5=CC(=CN=C5)Br

Specifications

Color White
Quantity 1 g
Merck Index 14,6496
Solubility Information Soluble in alcohol,chloroform,and acetone. Insoluble in water.
Formula Weight 484.39
Physical Form Powder
Chemical Name or Material Nicergoline
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 248-694-6
RTECSNumber KE6341000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.