Learn More
Nickel bis(trifluoromethylsulfonyl)imide
Description
Nickel(II) Bis(trifluoromethanesulfonyl)imide is used as Surface treatment, Synthesis and catalysis. It is also used as a ionic liquid in synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Green |
| Physical Form | Solid |
| Quantity | 5 g |
| Linear Formula | [(CF3SO2)2N]2Ni |
| UN Number | UN3261 |
| Solubility Information | It is soluble in water. |
| Formula Weight | 618.96 |
| Sensitivity | Hygroscopic |
| Chemical Name or Material | Nickel bis(trifluoromethylsulfonyl)imide |
Safety and Handling
| Hazard Category | H314-H317-H335-H350 |
| Hazard Statement | GHS H Statement H350-H314-H318-H317 May cause cancer. Causes severe skin burns and eye damage. Causes serious eye damage. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P260-P264b-P271-P272-P280g-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.